3-Ethoxy-4-methoxybenzonitrile 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A255775-10G A255775-25G A255775-500G A255775-5G A255775-100G
77014-070EA 9 USD
77014-070 77014-072 77014-074 77014-076 77683-777
3-Ethoxy-4-methoxybenzonitrile 98%, Ambeed.Inc
3-Ethoxy-4-methoxybenzonitrile

Precaución: For research use only.

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD02256160
CAS Number: 60758-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR