(S)-tert-Butyl 2-amino-3-phenylpropanoate 95%

Supplier: Ambeed
Warning

Synonyms: H-3-Phe-OtBu

A255760-1G A255760-250MG A255760-5G
77000-800EA 62.6 USD
77000-800 77000-802 77000-804
(S)-tert-Butyl 2-amino-3-phenylpropanoate 95%
(S)-tert-Butyl 2-amino-3-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.29 g/mol
MDL Number: MFCD00038270
CAS Number: 16874-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR