N-Boc-D-alaninol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol

A255717-100G A255717-10G A255717-1G A255717-25G A255717-5G
77000-790EA 67 USD
77000-790 77000-792 77000-794 77000-796 77000-798
N-Boc-D-alaninol 97%
N-Boc-D-alaninol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MDL Number: MFCD00235912
CAS Number: 106391-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR