5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-(trifluoromethyl)pyridin-2-ol

A255665-100MG A255665-10G A255665-1G A255665-250MG A255665-5G A255665-25G
77031-670EA 5 USD
77031-670 77031-672 77031-674 77031-676 77031-678 77872-265
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃NO
MW: 242.00 g/mol
MDL Number: MFCD12546486
CAS Number: 76041-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR