4-Fluoro-2-(trifluoromethyl)phenol 97%
Supplier: Ambeed
|
Warning
|
A255609-10G
A255609-1G
A255609-250MG
A255609-25G
A255609-5G
77000-780EA
34
USD
77000-780
77000-782
77000-784
77000-786
77000-788
4-Fluoro-2-(trifluoromethyl)phenol 97%
4-Fluoro-2-(trifluoromethyl)phenol
Precaución: for Research Use Only
Formula:
C₇H₄F₄O |
MDL Number:
MFCD00061290 CAS Number: 130047-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...