4-Bromo-2-formylthiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1,3-thiazole-2-carboxaldehyde

A255185-1G A255185-250MG A255185-25G A255185-5G
77031-654EA 21.22 USD
77031-654 77031-656 77031-658 77031-660
4-Bromo-2-formylthiazole 97%
4-Bromo-2-formylthiazole

Precaución: for Research Use Only

Formula: C₄H₂BrNOS
MW: 192.0338 g/mol
MDL Number: MFCD06660122
CAS Number: 167366-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR