1-Boc-3-Ethynylpiperidine 97%

Supplier: Ambeed
Warning

A255076-100MG A255076-10G A255076-1G A255076-250MG A255076-5G
77031-644EA 8 USD
77031-644 77031-646 77031-648 77031-650 77031-652
1-Boc-3-Ethynylpiperidine 97%
1-Boc-3-Ethynylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₂
MDL Number: MFCD10698159
CAS Number: 664362-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR