(2-Carboxyethyl)phenylphosphinic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3-(Hydroxy(phenyl)phosphoryl)propanoic acid, 3-Hydroxyphenylphosphinylpropanoic acid

A255047-1G A255047-25G A255047-500G A255047-5G
77000-772EA 5 USD
77000-772 77000-774 77000-776 77000-778
(2-Carboxyethyl)phenylphosphinic acid 97%
(2-Carboxyethyl)phenylphosphinic acid

Precaución: for Research Use Only

Formula: C₉H₁₁O₄P
MW: 214.16 g/mol
MDL Number: MFCD08704562
CAS Number: 14657-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR