Fmoc-D-4-fluorophe 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(4-F)-OH, Fmoc-p-fluoro-D-Phe-OH, N-Fmoc-4-fluoro-D-phenylalanine

A254601-1G A254601-25G A254601-5G
77000-744EA 17 USD
77000-744 77000-746 77000-748
Fmoc-D-4-fluorophe 95%
Fmoc-D-4-fluorophe

Precaución: for Research Use Only

Formula: C₂₄H₂₀FNO₄
MW: 405.425 g/mol
MDL Number: MFCD00235900
CAS Number: 177966-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR