Ethyl 4-bromo-2,3-dihydroxybenzoate 95%

Supplier: Ambeed
Warning

A254432-1G A254432-250MG
77014-062EA 401.03 USD
77014-062 77014-064
Ethyl 4-bromo-2,3-dihydroxybenzoate 95%
Ethyl 4-bromo-2,3-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₄
MW: 261.06 g/mol
MDL Number: MFCD27996707
CAS Number: 1312609-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR