6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid 98%

Supplier: Ambeed
Warning

A254273-10G A254273-250MG A254273-5G A254273-1G A254273-25G
77000-738EA 47 USD
77000-738 77000-740 77000-742 77601-046 77881-687
6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid 98%
6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₂
MDL Number: MFCD04114705
CAS Number: 189005-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR