Naphthol AS-E 97%

Supplier: Ambeed
Danger

Synonyms: Naphthol AS-E, N-(4-Chlorophenyl)-3-hydroxy-2-naphthamide, Azoic Coupling Component 10, 4'-Chloro-3-hydroxy-2-naphthanilide

A254192-25G A254192-100G A254192-500G
77031-628EA 7.43 USD
77031-628 77601-042 77601-044
Naphthol AS-E 97%
Naphthol AS-E

Precaución: for Research Use Only

Formula: C₁₇H₁₂ClNO₂
MW: 297.74 g/mol
MDL Number: MFCD00021639
CAS Number: 92-78-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR