4-Iodo-1-methylimidazole-5-carbaldehyde 95+%

Supplier: Ambeed
Danger

Synonyms: 4-Iodo-1-methyl-1H-imidazole-5-carbaldehyde

A253988-1G A253988-250MG
77014-058EA 335.25 USD
77014-058 77014-060
4-Iodo-1-methylimidazole-5-carbaldehyde 95+%
4-Iodo-1-methylimidazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅IN₂O
MW: 236.0105 g/mol
MDL Number: MFCD01632219
CAS Number: 189014-13-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR