3-Bromo-o-toluonitrile 98%

Supplier: Ambeed
Warning

A253869-100G A253869-10G A253869-1G A253869-25G A253869-5G
77031-618EA 211 USD
77031-618 77031-620 77031-622 77031-624 77031-626
3-Bromo-o-toluonitrile 98%
3-Bromo-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MDL Number: MFCD09025665
CAS Number: 52780-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR