[1-(Aminomethyl)cyclobutyl]methanol 98%

Supplier: Ambeed
Danger

Synonyms: (1-(Aminomethyl)cyclobutyl)methanol

A253744-1G A253744-250MG A253744-5G A253744-100MG
77031-612EA 165.51 USD
77031-612 77031-614 77031-616 77672-272
[1-(Aminomethyl)cyclobutyl]methanol 98%
[1-(Aminomethyl)cyclobutyl]methanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD09864389
CAS Number: 2041-56-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR