N-Cbz-n'-trityl-L-glutamine 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Benzyloxycarbonyl-Nδ-trityl-L-glutamine, Z-Gln(Trt)-OH

A253612-10G A253612-1G A253612-25G
77000-732EA 28 USD
77000-732 77000-734 77000-736
N-Cbz-n'-trityl-L-glutamine 97%
N-Cbz-n'-trityl-L-glutamine

Precaución: for Research Use Only

Formula: C₃₂H₃₀N₂O₅
MDL Number: MFCD00144845
CAS Number: 132388-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR