3-Bromo-2,6-dimethoxypyridine 98%

Supplier: Ambeed
Warning

A253599-1G A253599-25G A253599-5G
77031-604EA 9.55 USD
77031-604 77031-606 77031-608
3-Bromo-2,6-dimethoxypyridine 98%
3-Bromo-2,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MDL Number: MFCD08669938
CAS Number: 13445-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR