2-(Boc-aminomethyl)benzyl Alcohol 98%

Supplier: Ambeed
Warning

A253456-100MG A253456-250MG
77014-042EA 21.22 USD
77014-042 77014-044
2-(Boc-aminomethyl)benzyl Alcohol 98%
2-(Boc-aminomethyl)benzyl Alcohol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.30 g/mol
MDL Number: MFCD22418063
CAS Number: 1333114-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR