Pyrimidine-4(3H)-thione 96%

Supplier: Ambeed
Warning

A253453-1G A253453-250MG A253453-5G
76987-968EA 110 USD
76987-968 76987-970 77890-426
Pyrimidine-4(3H)-thione 96%
Pyrimidine-4(3H)-thione

Precaución: for Research Use Only

Formula: C₄H₄N₂S
MDL Number: MFCD18089671
CAS Number: 1450-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR