2,6-Diethylpyridine 95%

Supplier: Ambeed
Danger

A253159-100MG A253159-250MG A253159-1G A253159-5G
77031-594EA 15 USD
77031-594 77031-596 77668-064 77872-260
2,6-Diethylpyridine 95%
2,6-Diethylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD00049215
CAS Number: 935-28-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR