[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
[1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II), Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)
A253007-1G
A253007-5G
A253007-250MG
77000-728EA
28
USD
77000-728
77000-730
77672-266
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride 97%
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride
Precaución: for Research Use Only
Formula:
C₂₆H₂₄Cl₂P₂Pd |
MDL Number:
MFCD00015702 CAS Number: 19978-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...