Ethyl 2-((4-methoxyphenyl)amino)-2-oxoacetate 95+%

Supplier: Ambeed
Danger

A252991-1G A252991-250MG A252991-5G
76987-950EA 35.01 USD
76987-950 76987-952 76987-954
Ethyl 2-((4-methoxyphenyl)amino)-2-oxoacetate 95+%
Ethyl 2-((4-methoxyphenyl)amino)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.22 g/mol
MDL Number: MFCD00157580
CAS Number: 18522-99-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR