3,5-Dimethylisonicotinonitrile 98%

Supplier: Ambeed
Danger

A252928-100MG A252928-250MG
76987-938EA 85 USD
76987-938 76987-940
3,5-Dimethylisonicotinonitrile 98%
3,5-Dimethylisonicotinonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD11052634
CAS Number: 7584-08-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR