4-(4-Nitrophenyl)oxazole 95%

Supplier: Ambeed
Warning

A252906-250MG A252906-5G A252906-1G
77000-720EA 11 USD
77000-720 77000-722 77672-264
4-(4-Nitrophenyl)oxazole 95%
4-(4-Nitrophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD09032979
CAS Number: 13382-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR