4,6-Dimethyl-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A252795-1G A252795-250MG
77000-718EA 44 USD
77000-718 77881-681
4,6-Dimethyl-1H-indole-2-carboxylic acid 98%
4,6-Dimethyl-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD02664431
CAS Number: 383132-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR