2-Hydrazinopyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydrazinylpyrimidine

A252645-10G A252645-1G A252645-250MG A252645-5G
77031-574EA 30 USD
77031-574 77031-576 77031-578 77672-262
2-Hydrazinopyrimidine 97%
2-Hydrazinopyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD01249337
CAS Number: 7504-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR