BAY 11-7085 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(4-tert-Butylphenylsulphonyl)acrylonitrile, (E)-3-((4-(tert-Butyl)phenyl)sulphonyl)acrylonitrile

A252603-10MG A252603-1MG A252603-25MG A252603-50MG A252603-5MG
76987-922EA 76.39 USD
76987-922 76987-924 76987-926 76987-928 76987-930
BAY 11-7085 98%
BAY 11-7085

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₂S
MW: 249.328 g/mol
MDL Number: MFCD01862602
CAS Number: 196309-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR