4-(4'-Fluorobenzyloxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Fluorobenzyl)oxy)phenyl)boronic acid

A252384-1G A252384-250MG A252384-5G
77014-014EA 39 USD
77014-014 77014-016 77014-018
4-(4'-Fluorobenzyloxy)phenylboronic acid 95%
4-(4'-Fluorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD07784403
CAS Number: 871125-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR