6-Methoxypurine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-9H-purine

A252196-1G A252196-250MG A252196-5G
77000-702EA 33.95 USD
77000-702 77000-704 77000-706
6-Methoxypurine 97%
6-Methoxypurine

Precaución: for Research Use Only

Formula: C₆H₆N₄O
MW: 150.1400 g/mol
MDL Number: MFCD00005574
CAS Number: 1074-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR