4-Amino-3,5-dichloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

A251906-100G A251906-10G A251906-25G A251906-500G A251906-5G
76987-908EA 30 USD
76987-908 76987-910 76987-912 76987-914 76987-916
4-Amino-3,5-dichloropyridine 98%
4-Amino-3,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD00125023
CAS Number: 22889-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR