Methyl 2-(4-bromophenyl)-2-oxoacetate 97%

Supplier: Ambeed
Warning

A251848-100MG A251848-1G
77014-012EA 21.22 USD
77014-012 77612-430
Methyl 2-(4-bromophenyl)-2-oxoacetate 97%
Methyl 2-(4-bromophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.05 g/mol
MDL Number: MFCD00193630
CAS Number: 57699-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR