3-Amino-4-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A251846-100MG A251846-1G A251846-250MG
77031-538EA 12 USD
77031-538 77031-540 77031-542
3-Amino-4-methoxybenzonitrile 97%
3-Amino-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD09745274
CAS Number: 60979-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR