2,4-Dihydroxypyrimidine-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isoorotic acid, Uracil-5-carboxylic acid

A251801-100G A251801-10G A251801-1G A251801-25G A251801-5G A251801-50MG A251801-500G
77000-690EA 86 USD
77000-690 77000-692 77000-694 77000-696 77000-698 77000-700 77672-252 77881-678
2,4-Dihydroxypyrimidine-5-carboxylic acid 98%
2,4-Dihydroxypyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
MDL Number: MFCD00006023
CAS Number: 23945-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR