4-(4-Chlorobenzyloxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Chlorobenzyl)oxy)phenyl)boronic acid

A251655-100MG A251655-1G A251655-250MG A251655-5G
77014-004EA 23.34 USD
77014-004 77014-006 77014-008 77014-010
4-(4-Chlorobenzyloxy)phenylboronic acid 95%
4-(4-Chlorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.5000 g/mol
MDL Number: MFCD07784349
CAS Number: 870778-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR