4-Bromo-7-methyl-2,3-dihydro-1H-inden-1-one 98%

Supplier: Ambeed
Warning

A251591-100MG A251591-250MG
76987-900EA 47.75 USD
76987-900 76987-902
4-Bromo-7-methyl-2,3-dihydro-1H-inden-1-one 98%
4-Bromo-7-methyl-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrO
MW: 225.08 g/mol
MDL Number: MFCD17018679
CAS Number: 90772-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR