Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate 99+%

Supplier: Ambeed
Danger

Synonyms: Sebacic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester, Bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(2,2,6,6-tetramethyl-4-piperidyl) Ester

A251447-100G A251447-25G A251447-500G
77000-676EA 17 USD
77000-676 77000-678 77000-680
Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate 99+%
Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate

Precaución: for Research Use Only

Formula: C₂₈H₅₂N₂O₄
MW: 480.73 g/mol
MDL Number: MFCD00134709
CAS Number: 52829-07-9
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR