2-(Methylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile 95%

Supplier: Ambeed
Warning

A251021-100MG A251021-1G A251021-250MG
76987-894EA 53.05 USD
76987-894 76987-896 76987-898
2-(Methylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile 95%
2-(Methylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₈BN₃O₂
MW: 259.11 g/mol
MDL Number: MFCD16995277
CAS Number: 1346809-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR