4-(4-Pyridyl)-3-butyn-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yl)but-3-yn-1-ol

A250890-1G A250890-250MG A250890-5G
77013-984EA 61 USD
77013-984 77013-986 77890-417
4-(4-Pyridyl)-3-butyn-1-ol 95%
4-(4-Pyridyl)-3-butyn-1-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
MDL Number: MFCD18384538
CAS Number: 192643-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR