2-Bromo-5-fluoro-4-methylbenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A250503-1G
A250503-250MG
A250503-5G
76987-874EA
25
USD
76987-874
76987-876
77890-416
2-Bromo-5-fluoro-4-methylbenzoic acid 97%
2-Bromo-5-fluoro-4-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ |
MDL Number:
MFCD09263436 CAS Number: 1003709-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...