2-(Hexahydro-1H-pyrrolizin-7a-yl)acetonitrile 97%
Supplier: Ambeed
|
Warning
|
A250461-1G
A250461-250MG
A250461-5G
77013-972EA
50.93
USD
77013-972
77013-974
77013-976
2-(Hexahydro-1H-pyrrolizin-7a-yl)acetonitrile 97%
2-(Hexahydro-1H-pyrrolizin-7a-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₉H₁₄N₂ MW: 150.22 g/mol |
MDL Number:
MFCD19704706 CAS Number: 78449-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...