2-Cyano-6-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Cyano-6-fluorophenyl)boronic acid

A250222-100MG A250222-250MG A250222-5G
76987-866EA 31.83 USD
76987-866 76987-868 77683-748
2-Cyano-6-fluorophenylboronic acid 95%
2-Cyano-6-fluorophenylboronic acid

Precaución: For research use only.

Formula: C₇H₅BFNO₂
MW: 164.9295 g/mol
MDL Number: MFCD09033623
CAS Number: 656235-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR