tert-Butyl 2-methyl-4-oxopyrrolidine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A249642-100MG
A249642-1G
A249642-250MG
76987-846EA
85.94
USD
76987-846
76987-848
76987-850
tert-Butyl 2-methyl-4-oxopyrrolidine-1-carboxylate 98%
tert-Butyl 2-methyl-4-oxopyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol |
MDL Number:
MFCD22376634 CAS Number: 362706-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...