Isoquinoline-4-carbonitrile 95%

Supplier: Ambeed
Warning

A249625-10G A249625-1G A249625-250MG A249625-5G
77013-958EA 210.06 USD
77013-958 77013-960 77013-962 77013-964
Isoquinoline-4-carbonitrile 95%
Isoquinoline-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.1700 g/mol
MDL Number: MFCD00957039
CAS Number: 34846-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR