2,2'-Dithiodi(benzoic acid) 96%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Dithiosalicylic acid, 2,2'-Dithiodibenzoic acid

A249546-1000G A249546-25G A249546-500G A249546-100G
77000-644EA 84 USD
77000-644 77000-646 77000-648 77568-420
2,2'-Dithiodi(benzoic acid) 96%
2,2'-Dithiodi(benzoic acid)

Precaución: For research use only.

Formula: C₁₄H₁₀O₄S₂
MW: 306.36 g/mol
MDL Number: MFCD00002465
CAS Number: 119-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR