5-Amino-4-chloro-2-methylphenol 96%

Supplier: Ambeed
Warning

A249372-1G A249372-250MG
77000-624EA 62.6 USD
77000-624 77000-626
5-Amino-4-chloro-2-methylphenol 96%
5-Amino-4-chloro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.60 g/mol
MDL Number: MFCD09744010
CAS Number: 110102-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR