2-((tert-Butoxycarbonyl)amino)-2-cyclobutylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-2-Cyclobutylglycine, Boc-DL-2-Cyclobutyl-Gly-OH

A249356-100MG A249356-250MG A249356-1G A249356-5G
77013-950EA 19 USD
77013-950 77013-952 77890-413 77890-414
2-((tert-Butoxycarbonyl)amino)-2-cyclobutylacetic acid 97%
2-((tert-Butoxycarbonyl)amino)-2-cyclobutylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD09264345
CAS Number: 811460-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR