5-Bromo-1-ethyl-1H-indole 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A248648-1G A248648-5G
76987-838EA 29 USD
76987-838 77683-744
5-Bromo-1-ethyl-1H-indole 97%, Ambeed.Inc
5-Bromo-1-ethyl-1H-indole

Precaución: For research use only.

Formula: C₁₀H₁₀BrN
MDL Number: MFCD04344211
CAS Number: 195253-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR