3-Aminobenzoic acid 97%

Supplier: Ambeed
Warning

A248547-1000G A248547-100G A248547-10G A248547-25G A248547-500G
77031-450EA 90.18 USD
77031-450 77031-452 77031-454 77031-456 77031-458
3-Aminobenzoic acid 97%
3-Aminobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00007795
CAS Number: 99-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR