2-Amino-4,5-dichlorobenzaldehyde 97%

Supplier: Ambeed
Warning

A248412-100MG A248412-250MG
77013-942EA 20 USD
77013-942 77013-944
2-Amino-4,5-dichlorobenzaldehyde 97%
2-Amino-4,5-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
MDL Number: MFCD03066184
CAS Number: 163458-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR