6-Bromo-4-methoxypyridin-2-amine 98%

Supplier: Ambeed
Warning

A248360-100MG A248360-250MG A248360-5G A248360-1G
76987-830EA 82.76 USD
76987-830 76987-832 77612-392 77683-742
6-Bromo-4-methoxypyridin-2-amine 98%
6-Bromo-4-methoxypyridin-2-amine

Precaución: For research use only.

Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
MDL Number: MFCD20040422
CAS Number: 1158786-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR